siRNA Details
| Gene Symbol: Sgip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763917.1 | XP_008762139.1 | 14 | 1385 | ||
| XM_008763918.1 | XP_008762140.1 | 13 | 1300 | |||
| XM_008763919.1 | XP_008762141.1 | 16 | 1412 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgip1| Gene Symbol: Sgip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763917.1 | XP_008762139.1 | 14 | 1385 | ||
| XM_008763918.1 | XP_008762140.1 | 13 | 1300 | |||
| XM_008763919.1 | XP_008762141.1 | 16 | 1412 | |||