siRNA Details
| Gene Symbol: Sgip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763917.1 | XP_008762139.1 | 18, 19 | 2153 | ||
| XM_008763918.1 | XP_008762140.1 | 17, 18 | 2068 | |||
| XM_008763919.1 | XP_008762141.1 | 20, 21 | 2180 | |||

Sgip1
Genomic Map