siRNA Details
| Gene Symbol: Sgip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763917.1 | XP_008762139.1 | 20 | 2415 | ||
| XM_008763918.1 | XP_008762140.1 | 19 | 2330 | |||
| XM_008763919.1 | XP_008762141.1 | 22 | 2442 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgip1| Gene Symbol: Sgip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763917.1 | XP_008762139.1 | 20 | 2415 | ||
| XM_008763918.1 | XP_008762140.1 | 19 | 2330 | |||
| XM_008763919.1 | XP_008762141.1 | 22 | 2442 | |||