siRNA Details
| Gene Symbol: C8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191759.1 | NP_001178688.1 | 2 | 185 | ||
| XM_006238495.2 | XP_006238557.1 | 3 | 505 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8b| Gene Symbol: C8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191759.1 | NP_001178688.1 | 2 | 185 | ||
| XM_006238495.2 | XP_006238557.1 | 3 | 505 | |||