siRNA Details
| Gene Symbol: C8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191759.1 | NP_001178688.1 | 12 | 1634 | ||
| XM_006238495.2 | XP_006238557.1 | 13 | 1954 | |||
| GenBank mRNA | BC090023.1 | - | 9 | 1159 | ||
| FQ211372.1 | - | 7 | 618 | |||
| U20194.1 | - | 4 | 288 | |||

C8b
Genomic Map