siRNA Details
| Gene Symbol: Akap17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730183.4 | XP_002730229.1 | 5 | 2481 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akap17b| Gene Symbol: Akap17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730183.4 | XP_002730229.1 | 5 | 2481 | ||