siRNA Details
| Gene Symbol: Zcchc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014065.2 | NP_001014087.1 | 5 | 935 | ||
| XM_006257448.2 | XP_006257510.1 | 5 | 934 | |||
| XM_006257449.2 | XP_006257511.1 | 5 | 945 | |||
| XM_006257451.2 | XP_006257513.1 | 5 | 986 | |||
| XM_006257452.2 | XP_006257514.1 | 4 | 917 | |||
| XM_008773415.1 | XP_008771637.1 | 4 | 921 | |||
| GenBank mRNA | BC089115.1 | - | 5 | 935 | 7319855 | |

Zcchc12
Genomic Map