siRNA Details
| Gene Symbol: Dock11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191760.1 | NP_001178689.1 | 22 | 2551 | ||
| XM_006257453.2 | XP_006257515.1 | 22 | 2754 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dock11| Gene Symbol: Dock11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191760.1 | NP_001178689.1 | 22 | 2551 | ||
| XM_006257453.2 | XP_006257515.1 | 22 | 2754 | |||