siRNA Details
| Gene Symbol: Tmem255a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182822.3 | NP_877974.2 | 9 | 1053 | ||
| XM_006257479.2 | XP_006257541.1 | 8 | 984 | |||
| GenBank mRNA | AY327410.1 | - | 9 | 918 | ||
| CB610454.1 | - | 178 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem255a| Gene Symbol: Tmem255a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182822.3 | NP_877974.2 | 9 | 1053 | ||
| XM_006257479.2 | XP_006257541.1 | 8 | 984 | |||
| GenBank mRNA | AY327410.1 | - | 9 | 918 | ||
| CB610454.1 | - | 178 | ||||