siRNA Details
| Gene Symbol: Tmem255a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182822.3 | NP_877974.2 | 8 | 869 | ||
| XM_006257479.2 | XP_006257541.1 | 7 | 800 | |||
| GenBank mRNA | AY327410.1 | - | 8 | 734 | ||
| CB610453.1 | - | 203 | ||||
| CB610454.1 | - | 362 | ||||

Tmem255a
Genomic Map