siRNA Details
| Gene Symbol: Eps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009424.1 | NP_001009424.1 | 14, 15 | 1420 | ||
| GenBank mRNA | AB262963.1 | - | 14, 15 | 1276 | ||
| AY190609.1 | - | 14, 15 | 1420 | |||
| FQ223380.1 | - | 1408 | ||||

Eps15
Genomic Map