siRNA Details
| Gene Symbol: Eps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009424.1 | NP_001009424.1 | 15 | 1486 | ||
| GenBank mRNA | AB262963.1 | - | 15 | 1342 | ||
| AY190609.1 | - | 15 | 1486 | |||
| FQ223380.1 | - | 1474 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eps15| Gene Symbol: Eps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009424.1 | NP_001009424.1 | 15 | 1486 | ||
| GenBank mRNA | AB262963.1 | - | 15 | 1342 | ||
| AY190609.1 | - | 15 | 1486 | |||
| FQ223380.1 | - | 1474 | ||||