siRNA Details
Gene Symbol: Cc2d1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270984.1 | NP_001257913.1 | 17 | 2065 | ||
GenBank mRNA | FQ212419.1 | - | 8 | 2786 | ||
FQ222909.1 | - | 14 | 1667 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cc2d1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270984.1 | NP_001257913.1 | 17 | 2065 | ||
GenBank mRNA | FQ212419.1 | - | 8 | 2786 | ||
FQ222909.1 | - | 14 | 1667 |