siRNA Details
| Gene Symbol: Reck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107954.1 | NP_001101424.1 | 16 | 1909 | ||
| XM_006238068.2 | XP_006238130.1 | 16 | 1982 | |||
| XM_006238069.2 | XP_006238131.1 | 11 | 2627 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reck| Gene Symbol: Reck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107954.1 | NP_001101424.1 | 16 | 1909 | ||
| XM_006238068.2 | XP_006238130.1 | 16 | 1982 | |||
| XM_006238069.2 | XP_006238131.1 | 11 | 2627 | |||