siRNA Details
| Gene Symbol: Tmem8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238139.2 | XP_006238201.1 | 3 | 880 | ||
| XM_008763687.1 | XP_008761909.1 | 3 | 882 | |||
| XM_008763688.1 | XP_008761910.1 | 4 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem8b| Gene Symbol: Tmem8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238139.2 | XP_006238201.1 | 3 | 880 | ||
| XM_008763687.1 | XP_008761909.1 | 3 | 882 | |||
| XM_008763688.1 | XP_008761910.1 | 4 | 406 | |||