siRNA Details
| Gene Symbol: Dmbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107961.2 | NP_001101431.1 | 1, 2 | 175 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmbx1| Gene Symbol: Dmbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107961.2 | NP_001101431.1 | 1, 2 | 175 | ||