siRNA Details
| Gene Symbol: Hpdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014068.1 | NP_001014090.1 | 1 | 1180 | ||
| GenBank mRNA | BC083702.1 | - | 1 | 1180 | 7190108 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hpdl| Gene Symbol: Hpdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014068.1 | NP_001014090.1 | 1 | 1180 | ||
| GenBank mRNA | BC083702.1 | - | 1 | 1180 | 7190108 | |