siRNA Details
| Gene Symbol: Tmem53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107964.1 | NP_001101434.1 | 1 | 109 | ||
| XM_006238666.2 | XP_006238728.1 | 1 | 38 | |||
| GenBank mRNA | FQ219156.1 | - | 1 | 63 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem53| Gene Symbol: Tmem53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107964.1 | NP_001101434.1 | 1 | 109 | ||
| XM_006238666.2 | XP_006238728.1 | 1 | 38 | |||
| GenBank mRNA | FQ219156.1 | - | 1 | 63 | ||