siRNA Details
| Gene Symbol: Ago1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764025.1 | XP_008762247.1 | 10 | 1495 | ||
| GenBank mRNA | FQ234926.1 | - | 3 | 264 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ago1| Gene Symbol: Ago1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764025.1 | XP_008762247.1 | 10 | 1495 | ||
| GenBank mRNA | FQ234926.1 | - | 3 | 264 | ||