siRNA Details
Gene Symbol: Ago1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764025.1 | XP_008762247.1 | 15 | 2194 | ||
GenBank mRNA | FQ234926.1 | - | 8 | 963 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ago1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764025.1 | XP_008762247.1 | 15 | 2194 | ||
GenBank mRNA | FQ234926.1 | - | 8 | 963 |