siRNA Details
| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012104.1 | NP_001012104.1 | 6 | 625 | ||
| GenBank mRNA | BC082075.1 | - | 6 | 625 | 7133207 | |
| FQ214046.1 | - | 1 | 71 | |||
| FQ228905.1 | - | 6 | 657 | |||
| FQ229122.1 | - | 6 | 637 | |||

Ddost
Genomic Map