siRNA Details
| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012104.1 | NP_001012104.1 | 6 | 576 | ||
| GenBank mRNA | BC082075.1 | - | 6 | 576 | 7133207 | |
| FQ214046.1 | - | 1 | 22 | |||
| FQ228905.1 | - | 6 | 608 | |||
| FQ229122.1 | - | 6 | 588 | |||

Ddost
Genomic Map