siRNA Details
| Gene Symbol: Ubr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039026.1 | NP_001034115.1 | 49, 50 | 7469 | ||
| XM_006239202.2 | XP_006239264.1 | 49, 50 | 7520 | |||
| XM_006239206.2 | XP_006239268.1 | 49, 50 | 7520 | |||
| XM_006239207.2 | XP_006239269.1 | 49, 50 | 7520 | |||
| GenBank mRNA | AY850927.1 | - | 49, 50 | 7469 | ||

Ubr4
Genomic Map