siRNA Details
| Gene Symbol: Clstn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007092.1 | NP_001007093.1 | 5 | 913 | ||
| GenBank mRNA | AY569014.1 | - | 5 | 913 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clstn1| Gene Symbol: Clstn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007092.1 | NP_001007093.1 | 5 | 913 | ||
| GenBank mRNA | AY569014.1 | - | 5 | 913 | ||