siRNA Details
Gene Symbol: Errfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014071.1 | NP_001014093.1 | 4 | 914 | ||
XM_008764260.1 | XP_008762482.1 | 3 | 3662 | |||
XM_008764261.1 | XP_008762483.1 | 4 | 998 | |||
XM_008764262.1 | XP_008762484.1 | 3 | 3433 | |||
GenBank mRNA | BC083845.1 | - | 4 | 914 | 7121187 | |
X07266.1 | - | 4 | 917 |
