siRNA Details
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_233702.7 | XP_233702.4 | 11 | 1973 | ||
GenBank mRNA | BC098821.1 | - | 5 | 706 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_233702.7 | XP_233702.4 | 11 | 1973 | ||
GenBank mRNA | BC098821.1 | - | 5 | 706 |