siRNA Details
| Gene Symbol: Nphp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037650.2 | NP_001032739.2 | 4 | 770 | ||
| XM_006239464.2 | XP_006239526.1 | 4 | 8442 | |||
| XM_006239465.2 | XP_006239527.1 | 3 | 482 | |||
| XM_006239467.2 | XP_006239529.1 | 4 | 1827 | |||
| XM_008764358.1 | XP_008762580.1 | 5 | 1957 | |||
| XM_008764359.1 | XP_008762581.1 | 4 | 8442 | |||

Nphp4
Genomic Map