siRNA Details
| Gene Symbol: Noc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033897.1 | NP_001029069.1 | 13 | 1747 | ||
| XM_006239584.2 | XP_006239646.1 | 14 | 1661 | |||
| GenBank mRNA | BC101911.1 | - | 13 | 1747 | 7934070 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Noc2l| Gene Symbol: Noc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033897.1 | NP_001029069.1 | 13 | 1747 | ||
| XM_006239584.2 | XP_006239646.1 | 14 | 1661 | |||
| GenBank mRNA | BC101911.1 | - | 13 | 1747 | 7934070 | |