siRNA Details
| Gene Symbol: Dnajb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108004.1 | NP_001101474.1 | 4 | 743 | ||
| XM_006238070.2 | XP_006238132.1 | 4 | 776 | |||
| XM_006238071.2 | XP_006238133.1 | 3 | 1179 | |||
| XM_006238072.2 | XP_006238134.1 | 3 | 595 | |||
| GenBank mRNA | BC166524.1 | - | 3 | 592 | ||

Dnajb5
Genomic Map