siRNA Details
| Gene Symbol: Eml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108008.1 | NP_001101478.1 | 17, 18 | 1885 | ||
| XM_006239681.2 | XP_006239743.1 | 18, 19 | 2772 | |||
| XM_006239682.2 | XP_006239744.1 | 17, 18 | 2739 | |||

Eml4
Genomic Map