siRNA Details
| Gene Symbol: Plekhh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191770.1 | NP_001178699.1 | 12 | 2157 | ||
| XM_008764459.1 | XP_008762681.1 | 13 | 2586 | |||
| XM_008764460.1 | XP_008762682.1 | 13 | 2294 | |||
| XM_008764461.1 | XP_008762683.1 | 13 | 2286 | |||
| XM_008764462.1 | XP_008762684.1 | 12 | 2665 | |||
| XM_008764463.1 | XP_008762685.1 | 11 | 2091 | |||

Plekhh2
Genomic Map