siRNA Details
| Gene Symbol: Fam98a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014073.1 | NP_001014095.1 | 6 | 732 | ||
| GenBank mRNA | BC089217.1 | - | 6 | 732 | 7304253 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam98a| Gene Symbol: Fam98a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014073.1 | NP_001014095.1 | 6 | 732 | ||
| GenBank mRNA | BC089217.1 | - | 6 | 732 | 7304253 | |