siRNA Details
| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239834.2 | XP_006239896.1 | 13, 14 | 1493 | ||
| XM_006239836.2 | XP_006239898.2 | 13, 14 | 1487 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp512| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239834.2 | XP_006239896.1 | 13, 14 | 1493 | ||
| XM_006239836.2 | XP_006239898.2 | 13, 14 | 1487 | |||