siRNA Details
| Gene Symbol: Smc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108014.1 | NP_001101484.1 | 4 | 452 | ||
| XM_006239898.2 | XP_006239960.1 | 4 | 611 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smc6| Gene Symbol: Smc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108014.1 | NP_001101484.1 | 4 | 452 | ||
| XM_006239898.2 | XP_006239960.1 | 4 | 611 | |||