siRNA Details
| Gene Symbol: Smc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108014.1 | NP_001101484.1 | 2, 3 | 277 | ||
| XM_006239898.2 | XP_006239960.1 | 2, 3 | 436 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025695, AK220488, BC099955, AK088002, AK050341, and others.

Smc6
Genomic Map