siRNA Details
| Gene Symbol: Smc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108014.1 | NP_001101484.1 | 16 | 1974 | ||
| XM_006239898.2 | XP_006239960.1 | 16 | 2133 | |||
| GenBank mRNA | FQ230575.1 | - | 5 | 721 | ||
| FQ234676.1 | - | 5 | 720 | |||

Smc6
Genomic Map