siRNA Details
| Gene Symbol: Nol10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014076.1 | NP_001014098.1 | 16 | 1389 | ||
| GenBank mRNA | BC081954.1 | - | 16 | 1389 | 7122941 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nol10| Gene Symbol: Nol10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014076.1 | NP_001014098.1 | 16 | 1389 | ||
| GenBank mRNA | BC081954.1 | - | 16 | 1389 | 7122941 | |