siRNA Details
| Gene Symbol: Baz1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170568.1 | NP_001164039.1 | 23 | 4385 | ||
| GenBank mRNA | CO565747.1 | - | 1 | 381 | ||
| FM109160.1 | - | 1 | 36 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Baz1a| Gene Symbol: Baz1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170568.1 | NP_001164039.1 | 23 | 4385 | ||
| GenBank mRNA | CO565747.1 | - | 1 | 381 | ||
| FM109160.1 | - | 1 | 36 | |||