siRNA Details
| Gene Symbol: Cdkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025121.1 | NP_001020292.1 | 3 | 408 | ||
| XM_006240171.2 | XP_006240233.1 | 4 | 1526 | |||
| XM_006240172.2 | XP_006240234.1 | 4 | 673 | |||
| GenBank mRNA | BC081896.1 | - | 3 | 408 | 7110321 | |

Cdkl1
Genomic Map