siRNA Details
| Gene Symbol: Daam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108030.1 | NP_001101500.1 | 24 | 3217 | ||
| XM_006240211.2 | XP_006240273.1 | 26 | 7783 | |||
| XM_006240212.2 | XP_006240274.1 | 25 | 7753 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Daam1| Gene Symbol: Daam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108030.1 | NP_001101500.1 | 24 | 3217 | ||
| XM_006240211.2 | XP_006240273.1 | 26 | 7783 | |||
| XM_006240212.2 | XP_006240274.1 | 25 | 7753 | |||