siRNA Details
Gene Symbol: Daam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108030.1 | NP_001101500.1 | 6 | 811 | ||
XM_006240211.2 | XP_006240273.1 | 6 | 5047 | |||
XM_006240212.2 | XP_006240274.1 | 6 | 5047 |
