siRNA Details
| Gene Symbol: Snapc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108033.1 | NP_001101503.1 | 5 | 681 | ||
| GenBank mRNA | BC161969.1 | - | 5 | 700 | 9028949 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snapc1| Gene Symbol: Snapc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108033.1 | NP_001101503.1 | 5 | 681 | ||
| GenBank mRNA | BC161969.1 | - | 5 | 700 | 9028949 | |