siRNA Details
| Gene Symbol: Dcaf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100718.1 | NP_001094188.1 | 8 | 1057 | ||
| XM_006240279.2 | XP_006240341.1 | 8 | 859 | |||
| XM_008764782.1 | XP_008763004.1 | 8 | 912 | |||
| GenBank mRNA | BC079428.1 | - | 3 | 176 | ||

Dcaf5
Genomic Map