siRNA Details
| Gene Symbol: Smoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002835.1 | NP_001002835.1 | 7 | 635 | ||
| XM_006240284.2 | XP_006240346.1 | 7 | 941 | |||
| XM_006240285.2 | XP_006240347.1 | 7 | 941 | |||
| XM_008764785.1 | XP_008763007.1 | 8 | 1041 | |||
| XM_008764786.1 | XP_008763008.1 | 8 | 1008 | |||
| GenBank mRNA | FQ222878.1 | - | 7 | 892 | ||

Smoc1
Genomic Map