siRNA Details
| Gene Symbol: Entpd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199394.1 | NP_955426.1 | 3 | 392 | ||
| XM_006240343.2 | XP_006240405.1 | 4 | 1080 | |||
| XM_006240344.1 | XP_006240406.1 | 4 | 440 | |||
| XM_006240345.2 | XP_006240407.1 | 4 | 1055 | |||
| XM_006240346.2 | XP_006240408.1 | 3 | 539 | |||
| GenBank mRNA | BC062044.1 | - | 3 | 392 | 5597848 | |

Entpd5
Genomic Map