siRNA Details
| Gene Symbol: Mlh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108043.1 | NP_001101513.1 | 8 | 3835 | ||
| XM_006240310.2 | XP_006240372.1 | 8 | 4496 | |||
| XM_006240311.2 | XP_006240373.1 | 8 | 4191 | |||
| XM_006240312.2 | XP_006240374.1 | 7 | 4104 | |||

Mlh3
Genomic Map