siRNA Details
| Gene Symbol: Lrrc74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037783.2 | NP_001032872.2 | 11 | 1618 | ||
| GenBank mRNA | BC083569.1 | - | 11 | 1567 | ||
| BC098927.1 | - | 1630 | ||||
| BC127444.1 | - | 11 | 1609 | 7135116 | ||
| CR475527.1 | - | 2 | 187 | |||

Lrrc74
Genomic Map