siRNA Details
| Gene Symbol: Cipc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108044.2 | NP_001101514.2 | 4 | 1225 | ||
| XM_006240377.2 | XP_006240439.1 | 4 | 1151 | |||
| XM_006240378.2 | XP_006240440.1 | 4 | 1864 | |||
| XM_006240380.2 | XP_006240442.1 | 4 | 1287 | |||
| XM_008764796.1 | XP_008763018.1 | 5 | 1404 | |||

Cipc
Genomic Map