siRNA Details
| Gene Symbol: Tdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031657.1 | NP_001026827.1 | 13 | 1617 | ||
| XM_006240432.2 | XP_006240494.1 | 14 | 1962 | |||
| XM_006240434.2 | XP_006240496.1 | 14 | 1633 | |||
| XM_006240435.2 | XP_006240497.1 | 14 | 2216 | |||
| GenBank mRNA | BC098739.1 | - | 13 | 1617 | 7385935 | |

Tdp1
Genomic Map