siRNA Details
| Gene Symbol: Nrde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_234465.8 | XP_234465.5 | 11 | 3336 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrde2| Gene Symbol: Nrde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_234465.8 | XP_234465.5 | 11 | 3336 | ||